AC5372912142 Accela ChemBio Inc.

4-tert-Butyl-N-phenylaniline

  • MDL number MFCD00443884
  • Cas Number : 4496-49-5
  • Molecular Weight : 225.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019514&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY019514.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart