AC5372912155 Accela ChemBio Inc.

2-Methyl-3-(1-piperidinyl)benzoic Acid

  • MDL number MFCD06740060
  • Cas Number : 886501-22-0
  • Molecular Weight : 219.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-3-(1-piperidinyl)benzoic acid
Inchi Code 1S/C13H17NO2/c1-10-11(13(15)16)6-5-7-12(10)14-8-3-2-4-9-14/h5-7H,2-4,8-9H2,1H3,(H,15,16)
Inchi Key RZQWNANFHHDHHJ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. RT
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019610&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY019610.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart