AC5372912157 Accela ChemBio Inc.

alpha-(3-Methylsulfonylphenyl)benzylamine

  • MDL number MFCD09817373
  • Cas Number : 1016509-13-9
  • Molecular Weight : 261.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019622&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY019622.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart