AC5372912160 Accela ChemBio Inc.

2-Methyl-3-morpholinobenzoic Acid

  • MDL number MFCD06740122
  • Cas Number : 886501-40-2
  • Molecular Weight : 221.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019631&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY019631.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart