AC5372912186 Accela ChemBio Inc.

2-Boc-5-bromo-1,2,3,4-tetrahydroisoquinolin-8-amine

  • MDL number MFCD13195444
  • Cas Number : 1260763-55-0
  • Molecular Weight : 327.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019846&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY019846.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart