AC5372912187 Accela ChemBio Inc.

5-Methyl-7,8-dihydro-[1,3]dioxolo[4,5-g]isoquinoline

  • MDL number MFCD08166716
  • Cas Number : 17104-27-7
  • Molecular Weight : 189.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form solid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019847&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY019847.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart