AC5372912191 Accela ChemBio Inc.

5-Amino-1-methylpyridin-2(1H)-one Hydrochloride

  • MDL number MFCD12197225
  • Cas Number : 1137721-06-2
  • Molecular Weight : 160.60146

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019857&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY019857.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart