AC5372912193 Accela ChemBio Inc.

Methyl 3-(Ethylamino)propanoate

  • MDL number MFCD09035843
  • Cas Number : 3440-30-0
  • Molecular Weight : 131.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019881&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY019881.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart