AC5372912212 Accela ChemBio Inc.

1-Methyl-5-nitro-2(1H)-pyridinone

  • MDL number MFCD00955768
  • Cas Number : 32896-90-5
  • Molecular Weight : 154.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-methyl-5-nitro-2(1H)-pyridinone
Inchi Code 1S/C6H6N2O3/c1-7-4-5(8(10)11)2-3-6(7)9/h2-4H,1H3
Inchi Key FZYSOPYUSSFGAO-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. RT
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY020000&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY020000.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart