AC5372912230 Accela ChemBio Inc.

4-Hydroxy-6-iodo-2-quinolinone

  • MDL number MFCD15527322
  • Cas Number : 1260760-00-6
  • Molecular Weight : 287.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY020277&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY020277.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart