AC5372912235 Accela ChemBio Inc.

2,3'-Dibromo-4'-methoxyacetophenone

  • MDL number MFCD02646974
  • Cas Number : 6096-83-9
  • Molecular Weight : 307.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY020299&attname=GoodsCode

Safety Information

signalword Danger
hcodes GHS05, GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY020299.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart