AC5372912243 Accela ChemBio Inc.

Triethyl 4-Phosphonobutyrate

  • MDL number MFCD00015168
  • Cas Number : 2327-69-7
  • Molecular Weight : 252.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY020318&attname=GoodsCode

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY020318.pdf
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart