AC5372912245 Accela ChemBio Inc.

Methyl 3-Amino-3-(3-pyridyl)propionate

  • MDL number MFCD11200699
  • Cas Number : 129042-89-3
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY020335&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY020335.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart