AC5372912248 Accela ChemBio Inc.

Ethyl (Z)-3-(4-Chlorophenyl)-2-cyanoacrylate

  • MDL number MFCD00454124
  • Cas Number : 2169-68-8
  • Molecular Weight : 235.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl (2Z)-3-(4-chlorophenyl)-2-cyano-2-propenoate
Inchi Code 1S/C12H10ClNO2/c1-2-16-12(15)10(8-14)7-9-3-5-11(13)6-4-9/h3-7H,2H2,1H3/b10-7-
Inchi Key KLUDCHZOLVGLQF-YFHOEESVSA-N

Description

Purity 97%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY020364&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY020364.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart