AC5372912249 Accela ChemBio Inc.

3-(Methylsulfonyl)benzophenone

  • MDL number MFCD11545096
  • Cas Number : 21205-05-0
  • Molecular Weight : 260.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY020370&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY020370.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart