AC5372912250 Accela ChemBio Inc.

5-Amino-1-methyl-2-piperidinone

  • MDL number MFCD09971184
  • Cas Number : 90485-53-3
  • Molecular Weight : 128.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY020379&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY020379.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart