AC5372912254 Accela ChemBio Inc.

2-Amino-4-bromo-5-fluorophenol

  • MDL number MFCD11101429
  • Cas Number : 1016234-89-1
  • Molecular Weight : 206.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form solid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY020499&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY020499.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart