AC5372912270 Accela ChemBio Inc.

(R)-4-Amino-6-fluorochromane Hydrochloride

  • MDL number MFCD15474862
  • Cas Number : 911826-09-0
  • Molecular Weight : 203.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY020642&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY020642.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart