AC5372912279 Accela ChemBio Inc.

4-Bromo-1H-indazol-3(2H)-one

  • MDL number MFCD13176950
  • Cas Number : 864845-15-8
  • Molecular Weight : 213.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-1,2-dihydro-3H-indazol-3-one
Inchi Code 1S/C7H5BrN2O/c8-4-2-1-3-5-6(4)7(11)10-9-5/h1-3H,(H2,9,10,11)
Inchi Key AOSWBTIWUXRHRT-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. +4?
Physical Form Yellow to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY020829&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY020829.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart