AC5372912289 Accela ChemBio Inc.

Ethyl 4-Chloro-2-indolecarboxylate

  • MDL number MFCD09264531
  • Cas Number : 53590-46-8
  • Molecular Weight : 223.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY020909&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY020909.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart