AC5372912298 Accela ChemBio Inc.

2-Chloro-4-ethylpyrimidine

  • MDL number MFCD10699651
  • Cas Number : 188707-99-5
  • Molecular Weight : 142.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021045&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY021045.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart