AC5372912306 Accela ChemBio Inc.

2-(2-Bromoethyl)oxirane

  • MDL number MFCD11040246
  • Cas Number : 13287-42-8
  • Molecular Weight : 151

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021183&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY021183.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart