AC5372912309 Accela ChemBio Inc.

1-Boc-3-(2,3-dichlorophenyl)piperazine

  • MDL number MFCD08686118
  • Cas Number : 886769-00-2
  • Molecular Weight : 331.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-(2,3-dichlorophenyl)-1-piperazinecarboxylate
Inchi Code 1S/C15H20Cl2N2O2/c1-15(2,3)21-14(20)19-8-7-18-12(9-19)10-5-4-6-11(16)13(10)17/h4-6,12,18H,7-9H2,1-3H3
Inchi Key UQFCGECXOICQHY-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021227&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY021227.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart