AC5372912316 Accela ChemBio Inc.

N-Ethyl-4-(dimethylamino)benzylamine

  • MDL number MFCD00465457
  • Cas Number : 313552-99-7
  • Molecular Weight : 178.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-[(ethylamino)methyl]-N,N-dimethylaniline
Inchi Code 1S/C11H18N2/c1-4-12-9-10-5-7-11(8-6-10)13(2)3/h5-8,12H,4,9H2,1-3H3
Inchi Key JPQUAUQJXYRDKM-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form Colorless to brown liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021252&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY021252.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart