AC5372912319 Accela ChemBio Inc.

N-Methyl-N-(2-pyridyl)glycine Methyl Ester

  • MDL number MFCD16151670
  • Cas Number : 1250236-70-4
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021379&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY021379.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart