AC5372912320 Accela ChemBio Inc.

7-Aminopyrazolo[1,5-a]pyrimidine-6-carbonitrile

  • MDL number MFCD00104599
  • Cas Number : 89975-57-5
  • Molecular Weight : 159.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021426&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY021426.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart