AC5372912325 Accela ChemBio Inc.

5-[2-(Cbz-amino)ethyl]-1H-tetrazole

  • MDL number MFCD22574988
  • Cas Number : 33841-54-2
  • Molecular Weight : 247.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name benzyl 2-(1H-tetraazol-5-yl)ethylcarbamate
Inchi Code 1S/C11H13N5O2/c17-11(12-7-6-10-13-15-16-14-10)18-8-9-4-2-1-3-5-9/h1-5H,6-8H2,(H,12,17)(H,13,14,15,16)
Inchi Key UJFKSVINPQBNQQ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. RT
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021588&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY021588.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart