AC5372912326 Accela ChemBio Inc.

2-(Dimethylamino)benzonitrile

  • MDL number MFCD01313389
  • Cas Number : 20925-24-0
  • Molecular Weight : 146.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021589&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY021589.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart