AC5388700947 Accela ChemBio Inc.

cis-3-Amino-1-hydroxy-4-methyl-2-pyrrolidinone Acetate

  • MDL number MFCD23381097
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022353&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY022353.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart