AC5388701027 Accela ChemBio Inc.

N-Ethyl-4-aminobenzylamine

  • MDL number MFCD09965833
  • Cas Number : 1019115-71-9
  • Molecular Weight : 150.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY003699&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY003699.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart