AC5388701040 Accela ChemBio Inc.

1-(4-Chlorophenyl)-3-(dimethylamino)propan-1-one Hydrochloride

  • MDL number MFCD00084951
  • Cas Number : 1798-83-0
  • Molecular Weight : 248.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY003981&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY003981.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart