AC5388701089 Accela ChemBio Inc.

D-N-Benzylserine Methyl Ester

  • MDL number MFCD12198843
  • Cas Number : 131110-76-4
  • Molecular Weight : 209.2417

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005134&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY005134.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart