AC5388701103 Accela ChemBio Inc.

Methyl 3-Aminopyridine-2-carboxylate

  • MDL number MFCD08695602
  • Cas Number : 36052-27-4
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005552&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY005552.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart