AC5388701155 Accela ChemBio Inc.

2-(Methylsulfonyl)-4-morpholinobenzaldehyde

  • MDL number MFCD13191595
  • Cas Number : 1197193-29-5
  • Molecular Weight : 269.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form Yellow to brown powder or solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006889&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY006889.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart