AC5388701188 Accela ChemBio Inc.

2-Amino-2-(4-pyridyl)ethanol Dihydrochloride

  • MDL number MFCD15474985
  • Cas Number : 1220039-63-3
  • Molecular Weight : 211.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY007453&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY007453.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart