AC5388701213 Accela ChemBio Inc.

Piperidine-2,6-dicarboxylic Acid

  • MDL number MFCD00986614
  • Cas Number : 499-82-1
  • Molecular Weight : 173.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form White to off white solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008131&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY008131.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart