AC5388701248 Accela ChemBio Inc.

2-(2-Imidazolyl)ethanol

  • MDL number MFCD06655989
  • Cas Number : 51036-79-4
  • Molecular Weight : 112.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY009459&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY009459.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart