AC5388701265 Accela ChemBio Inc.

Methyl 2-(4-Benzylpiperazino)-5-fluorobenzoate

  • MDL number MFCD18070990
  • Cas Number : 1256633-30-3
  • Molecular Weight : 328.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011015&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY011015.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart