AC5388701277 Accela ChemBio Inc.

4-Amino-6-isopropyl-2-methylpyrimidine

  • MDL number MFCD09909804
  • Cas Number : 95206-97-6
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011642&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY011642.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart