AC5388701279 Accela ChemBio Inc.

2-Chloro-1-(4-pyridinyl)ethanone Hydrochloride

  • MDL number MFCD08692051
  • Cas Number : 25260-36-0
  • Molecular Weight : 192.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011676&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY011676.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart