AC5388701286 Accela ChemBio Inc.

1-Benzyl-4-iodoimidazole

  • MDL number MFCD06412564
  • Cas Number : 536760-32-4
  • Molecular Weight : 284.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011828&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY011828.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart