AC5388701289 Accela ChemBio Inc.

Ethyl (R)-N-Cbz-3-pyrrolidinecarboxylate

  • MDL number MFCD11053571
  • Cas Number : 1263078-10-9
  • Molecular Weight : 277.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011942&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY011942.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart