AC5388701305 Accela ChemBio Inc.

2-[1-(Boc-amino)cyclopropyl]pyridine

  • MDL number MFCD18651687
  • Cas Number : 873221-80-8
  • Molecular Weight : 234.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012363&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY012363.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart