AC5388701306 Accela ChemBio Inc.

5-(4-Bromophenyl)-1-methylimidazole

  • MDL number MFCD18375271
  • Cas Number : 136350-71-5
  • Molecular Weight : 237.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012402&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012402.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart