AC5388701311 Accela ChemBio Inc.

Tetrahydrofuran-2-carboxamide

  • MDL number MFCD03929553
  • Cas Number : 91470-28-9
  • Molecular Weight : 115.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012530&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012530.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart