AC5388701316 Accela ChemBio Inc.

4-Bromo-2,6-dinitrobenzoic Acid

  • MDL number MFCD06410771
  • Cas Number : 95192-56-6
  • Molecular Weight : 291.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013028&attname=GoodsCode

Safety Information

signalword Warning
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY013028.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart