AC5388701328 Accela ChemBio Inc.

Methyl (S)-1-(Ethoxycarbonyl)pyrrolidine-2-carboxylate

  • MDL number MFCD06410843
  • Cas Number : 93423-88-2
  • Molecular Weight : 201.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013368&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY013368.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart