AC5388701375 Accela ChemBio Inc.

Ethyl 3-Nitropyridine-2-carboxylate

  • MDL number MFCD16621695
  • Cas Number : 229343-13-9
  • Molecular Weight : 196.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014816&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY014816.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart