AC5388701392 Accela ChemBio Inc.

2-Amino-6-fluoro-3-methylbenzoic Acid

  • MDL number MFCD06409108
  • Cas Number : 874804-26-9
  • Molecular Weight : 169.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015359&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015359.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart