AC5388701406 Accela ChemBio Inc.

1-(4-Bromophenyl)-4-oxocyclohexanecarboxylic Acid

  • MDL number MFCD06410579
  • Cas Number : 887978-75-8
  • Molecular Weight : 297.14452

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015845&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015845.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart